5-Acetyl-2,4-dimethylthiazole ≥98.0%

Supplier: TCI America

Synonyms: 2,4-Dimethyl-5-acetylthiazole, 1-(2,4-Dimethylthiazol-5-yl)ethanone, 1-(2,4-Dimethylthiazol-5-yl)ethan-1-one

A1215-25G A1215-10G
TCA1215-25GEA 97.49 CAD
TCA1215-25G TCA1215-010G
5-Acetyl-2,4-dimethylthiazole ≥98.0%
5-Acetyl-2,4-dimethylthiazole

CAS Number: 38205-60-6
MDL Number: MFCD00005331
Molecular Formula: C7H9NOS
Molecular Weight: 155.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 104
Specific Gravity (20/20): 1.15

Formula: C₇H₉NOS
MW: 155.22 g/mol
Boiling Pt: 230 °C
Density: 1.15 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00005331
CAS Number: 38205-60-6
EINECS: 253-826-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9aaf7eec4a79a17bb154dae4248f9adc16f8fe35&vid=9aaf7eec4a79a17bb154dae4248f9adc16f8fe35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR