2-Amino-4-hydroxy-6-methylpyrimidine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A1204-25G
A1204-250G
TCA1204-025GEA
55.3
CAD
TCA1204-025G
TCA1204-250G
2-Amino-4-hydroxy-6-methylpyrimidine ≥98.0% (by HPLC, titration analysis)
2-Amino-4-hydroxy-6-methylpyrimidine
CAS Number: 3977-29-5
MDL Number: MFCD00006095
Molecular Formula: C5H7N3O
Molecular Weight: 125.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 2-Amino-6-methyl-4-pyrimidinol
Formula:
C₅H₇N₃O |
MDL Number:
MFCD00006095 CAS Number: 3977-29-5 EINECS: 223-612-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...