5-Bromopentyl acetate ≥95.0%

Supplier: TCI America

A1195-5G A1195-25G A1195-100G
TCA1195-005GEA 37.92 CAD
TCA1195-005G TCA1195-025G TCA1195-100G
5-Bromopentyl acetate ≥95.0%
5-Bromopentyl acetate

CAS Number: 15848-22-3
MDL Number: MFCD00039204
Molecular Formula: C7H13BrO2
Molecular Weight: 209.08
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 113
Specific Gravity (20/20): 1.31


Other Product Names / Synonyms: Acetic Acid 5-Bromopentyl Ester, 5-Bromoamyl Acetate, Acetic Acid 5-Bromoamyl Ester

Formula: C₇H₁₃BrO₂
Boiling Pt: 113 °C
Density: 1.31 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00039204
CAS Number: 15848-22-3
EINECS: 239-961-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9d6f203e1ebb4e09866be1effbe3848201600b5&vid=b9d6f203e1ebb4e09866be1effbe3848201600b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR