4-Amino-3-hydroxybenzoic acid ≥98.0%

Supplier: TCI America
Warning

A1194-25G A1194-5G
TCA1194-25GEA 221.2 CAD
TCA1194-25G TCA1194-005G
4-Amino-3-hydroxybenzoic acid ≥98.0%
4-Amino-3-hydroxybenzoic acid

CAS Number: 2374-03-0
MDL Number: MFCD00017094
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₇H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00017094
CAS Number: 2374-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c27984f272655857dda75a7dc2550fc27d60628&vid=16cc7ac7ffc1ca3ffd2dab58d7f6705288778fc9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR