1-Amino-4-methylpiperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1192-500ML A1192-25ML
TCA1192-500MLEA 1804.36 CAD
TCA1192-500ML TCA1192-025ML
1-Amino-4-methylpiperazine ≥98.0% (by GC, titration analysis)
1-Amino-4-methylpiperazine

CAS Number: 6928-85-4
MDL Number: MFCD00006154
Molecular Formula: C5H13N3
Molecular Weight: 115.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 103
Specific Gravity (20/20): 0.96

Formula: C₅H₁₃N₃
Boiling Pt: 175 °C
Density: 0.96 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00006154
CAS Number: 6928-85-4
EINECS: 230-053-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac01cf750bd961084a66fee9faf078a84ad1a785&vid=ac01cf750bd961084a66fee9faf078a84ad1a785&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR