4-Chloro-3-sulfamoylbenzoic acid ≥98.0%
Supplier: TCI America
A1189-25G
A1189-500G
TCA1189-025GEA
34.76
CAD
TCA1189-025G
TCA1189-500G
4-Chloro-3-sulfamoylbenzoic acid ≥98.0%
4-Chloro-3-sulphamoylbenzoic acid
CAS Number: 1205-30-7
MDL Number: MFCD00012375
Molecular Formula: C7H6ClNO4S
Molecular Weight: 235.64
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 261
Other Product Names / Synonyms: 3-(Aminosulfonyl)-4-chlorobenzoic Acid
Formula:
C₇H₆ClNO₄S MW: 235.65 g/mol Melting Pt: 261 °C |
MDL Number:
MFCD00012375 CAS Number: 1205-30-7 EINECS: 214-882-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...