2-Acetyl-5-bromothiophene ≥98.0%

Supplier: TCI America

Synonyms: 1-(5-Bromo-2-thienyl)-1-ethanone

A1183-5G A1183-25G
TCA1183-5GEA 42.66 CAD
TCA1183-5G TCA1183-25G
2-Acetyl-5-bromothiophene ≥98.0%
2-Acetyl-5-bromothiophene

CAS Number: 5370-25-2
MDL Number: MFCD00014528
Molecular Formula: C6H5BrOS
Molecular Weight: 205.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 103
Melting point (°C): 95

Formula: C₆H₅BrOS
Boiling Pt: 103 °C
Melting Pt: 95 °C
MDL Number: MFCD00014528
CAS Number: 5370-25-2
EINECS: 226-363-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3df7a21220f6c6b04a7ac65ce7f6f0de7c5557b7&vid=3df7a21220f6c6b04a7ac65ce7f6f0de7c5557b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR