3-Acetyl-2,5-dichlorothiophene ≥98.0%

Supplier: TCI America

Synonyms: 1-(2,5-Dichloro-3-thienyl)-1-ethanone

A1182-25G A1182-5G
TCA1182-25GEA 368.14 CAD
TCA1182-25G TCA1182-5G
3-Acetyl-2,5-dichlorothiophene ≥98.0%
3-Acetyl-2,5-dichlorothiophene

CAS Number: 36157-40-1
MDL Number: MFCD00014522
Molecular Formula: C6H4Cl2OS
Molecular Weight: 195.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 122
Melting point (°C): 38
Flash Point (°C): 113

Formula: C₆H₄Cl₂OS
MW: 195.07 g/mol
Boiling Pt: 122 °C
Melting Pt: 38 °C
Flash Pt: 113 °C
MDL Number: MFCD00014522
CAS Number: 36157-40-1
EINECS: 252-893-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7e15eef51142d38114688d0bfd928e0ab5c6d4f&vid=f7e15eef51142d38114688d0bfd928e0ab5c6d4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR