4-Fluoro-3-(trifluoromethyl)aniline ≥98.0%

Supplier: TCI America
Warning

A1164-5G A1164-25G
TCA1164-005GEA 44.24 CAD
TCA1164-005G TCA1164-025G
4-Fluoro-3-(trifluoromethyl)aniline ≥98.0%
4-Fluoro-3-(trifluoromethyl)aniline

CAS Number: 2357-47-3
MDL Number: MFCD00007834
Molecular Formula: C7H5F4N
Molecular Weight: 179.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 92
Specific Gravity (20/20): 1.41


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-3-trifluoromethylaniline

Formula: C₇H₅F₄N
Boiling Pt: 208 °C
Density: 1.41 (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007834
CAS Number: 2357-47-3
EINECS: 219-095-7
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5905932401ceacbef81fc500a3306cb252f0f0f5&vid=5905932401ceacbef81fc500a3306cb252f0f0f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR