4-Fluoro-2-(trifluoromethyl)aniline ≥97.0%

Supplier: TCI America

Synonyms: α,α,α,4-Tetrafluoro-o-toluidine, 2-Amino-alpha,alpha,alpha,5-tetrafluorotoluene, 2-Amino-5-fluorobenzotrifluoride, 2-Amino-α,α,α,5-tetrafluorotoluene

A1158-25G A1158-5G
TCA1158-25GEA 294.2 CAD
TCA1158-25G TCA1158-005G
4-Fluoro-2-(trifluoromethyl)aniline ≥97.0%
4-Fluoro-2-(trifluoromethyl)aniline

CAS Number: 393-39-5
MDL Number: MFCD00007831
Molecular Formula: C7H5F4N
Molecular Weight: 179.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 72
Specific Gravity (20/20): 1.41


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-2-trifluoromethylaniline

Formula: C₇H₅F₄N
MW: 179.12 g/mol
Boiling Pt: 90 °C
Density: 1.41 (20 °C)
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007831
CAS Number: 393-39-5
EINECS: 206-886-7
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01afc8ba9d70d8b47261c9a01b76262c56bc31f5&vid=01afc8ba9d70d8b47261c9a01b76262c56bc31f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR