5-Amino-2-chloropyridine ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Chloro-5-pyridylamine, 6-Chloropyridin-3-amine, 2-Chloropyridin-5-amine

A1155-1G A1155-5G
TCA1155-001GEA 41.08 CAD
TCA1155-001G TCA1155-005G
5-Amino-2-chloropyridine ≥98.0%
5-Amino-2-chloropyridine

CAS Number: 5350-93-6
MDL Number: MFCD00006243
Molecular Formula: C5H5ClN2
Molecular Weight: 128.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 83

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 225
Melting Pt: 83
MDL Number: MFCD00006243
CAS Number: 5350-93-6
EINECS: 226-322-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e47c157992c582e09b03eab8f5245a79f21649e&vid=9e47c157992c582e09b03eab8f5245a79f21649e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR