2-Phenoxyaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A1142-25G
A1142-500G
TCA1142-025GEA
53.72
CAD
TCA1142-025G
TCA1142-500G
2-Phenoxyaniline ≥98.0% (by GC, titration analysis)
2-Phenoxyaniline
CAS Number: 2688-84-8
MDL Number: MFCD00035765
Molecular Formula: C12H11NO
Molecular Weight: 185.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 187
Melting point (°C): 46
Flash Point (°C): 110
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Aminophenyl Phenyl Ether, 2-Phenoxyaniline
Formula:
C₁₂H₁₁NO Boiling Pt: 187 °C Melting Pt: 46 °C Flash Pt: 110 °C |
MDL Number:
MFCD00035765 CAS Number: 2688-84-8 EINECS: 220-254-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...