4-Aminobenzophenone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1140-5G A1140-25G
TCA1140-005GEA 56.25 CAD
TCA1140-005G TCA1140-025G
4-Aminobenzophenone ≥98.0% (by titrimetric analysis)
4-Aminobenzophenone

CAS Number: 1137-41-3
MDL Number: MFCD00007895
Molecular Formula: C13H11NO
Molecular Weight: 197.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 123

Formula: C₁₃H₁₁NO
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00007895
CAS Number: 1137-41-3
EINECS: 214-506-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ccea1fefb8f4b5be058608b047560536ee345dd&vid=7ccea1fefb8f4b5be058608b047560536ee345dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR