4-Amino-4H-1,2,4-triazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A1137-250G A1137-25G
TCA1137-250GEA 259.12 CAD
TCA1137-250G TCA1137-25G
4-Amino-4H-1,2,4-triazole ≥98.0% (by HPLC, titration analysis)
4-Amino-4H-1,2,4-triazole

CAS Number: 584-13-4
MDL Number: MFCD00003099
Molecular Formula: C2H4N4
Molecular Weight: 84.08
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 86

Formula: C₂H₄N₄
Melting Pt: 86 °C
MDL Number: MFCD00003099
CAS Number: 584-13-4
EINECS: 209-533-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46b6df0bdc42a4ff0c89b7e2ab91d91a0dfaaa33&vid=46b6df0bdc42a4ff0c89b7e2ab91d91a0dfaaa33&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR