Methyl-p-anisate ≥99.0%
Supplier: TCI America
A1125-25G
A1125-500G
A1125-100G
TCA1125-025GEA
47.4
CAD
TCA1125-025G
TCA1125-500G
TCA1125-100G
Methyl-p-anisate ≥99.0%
Methyl p-anisate
CAS Number: 121-98-2
MDL Number: MFCD00008437
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 255
Melting point (°C): 49
Other Product Names / Synonyms: 4-Methoxybenzoic Acid Methyl Ester, p-Anisic Acid Methyl Ester, Methyl 4-Methoxybenzoate
Formula:
C₉H₁₀O₃ Boiling Pt: 255 °C Melting Pt: 49 °C |
MDL Number:
MFCD00008437 CAS Number: 121-98-2 EINECS: 204-513-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...