4-Chlorobutyl acetate ≥97.0%

Supplier: TCI America

A1098-250G A1098-25G
TCA1098-250GEA 398.16 CAD
TCA1098-250G TCA1098-25G
4-Chlorobutyl acetate ≥97.0%
4-Chlorobutyl acetate

CAS Number: 6962-92-1
MDL Number: MFCD00001013
Molecular Formula: C6H11ClO2
Molecular Weight: 150.60
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 87
Flash Point (°C): 64
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: Acetic Acid 4-Chlorobutyl Ester

Formula: C₆H₁₁ClO₂
Boiling Pt: 87 °C
Density: 1.08 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00001013
CAS Number: 6962-92-1
EINECS: 230-158-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7867f23316f33509f5e78a649512306343c46710&vid=7867f23316f33509f5e78a649512306343c46710&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR