α-Angelicalactone ≥98.0%
Supplier: TCI America
A1090-25G
A1090-100G
TCA1090-025GEA
93.22
CAD
TCA1090-025G
TCA1090-100G
α-Angelicalactone ≥98.0%
α-Angelicalactone
CAS Number: 591-12-8
MDL Number: MFCD00005375
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 56
Flash Point (°C): 68
Freezing point (°C): 16
Specific Gravity (20/20): 1.10
Product Categories: Bioscience, Hemiterpenes
Other Product Names / Synonyms: 4-Hydroxy-3-pentenoic Acid gamma-Lactone, 5-Methyl-2(3H)-furanone
Formula:
C₅H₆O₂ Boiling Pt: 56 °C Melting Pt: 0 °C Density: 1.10 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00005375 CAS Number: 591-12-8 EINECS: 209-701-8 Merck Index: 12,00686 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...