2-Amino-4-chloro-6-methylpyrimidine ≥98.0%

Supplier: TCI America

Synonyms: 4-Chloro-6-methylpyrimidin-2-amine

A1088-25G A1088-5G
TCA1088-025GEA 121.66 CAD
TCA1088-025G TCA1088-005G
2-Amino-4-chloro-6-methylpyrimidine ≥98.0%
2-Amino-4-chloro-6-methylpyrimidine

CAS Number: 5600-21-5
MDL Number: MFCD00006091
Molecular Formula: C5H6ClN3
Molecular Weight: 143.57
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 185

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00006091
CAS Number: 5600-21-5
EINECS: 227-018-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfec93e9596ad6b99f2c56a4cf53766363b74123&vid=bfec93e9596ad6b99f2c56a4cf53766363b74123&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR