2-Amino-4-chlorobenzothiazole ≥98.0% (by HPLC)

Supplier: TCI America

A1087-25G A1087-5G
TCA1087-25GEA 699.94 CAD
TCA1087-25G TCA1087-005G
2-Amino-4-chlorobenzothiazole ≥98.0% (by HPLC)
2-Amino-4-chlorobenzothiazole

CAS Number: 19952-47-7
MDL Number: MFCD00005791
Molecular Formula: C7H5ClN2S
Molecular Weight: 184.64
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 203

Formula: C₇H₅ClN₂S
MW: 184.65 g/mol
Melting Pt: 203 °C
MDL Number: MFCD00005791
CAS Number: 19952-47-7
EINECS: 243-439-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3fe4fbb5bf281dd2aa4207b99d96b2f78d37a4e&vid=a3fe4fbb5bf281dd2aa4207b99d96b2f78d37a4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR