4-Amino-1-benzylpiperidine ≥98.0%
Supplier: TCI America
Synonyms:
4-Amino-N-benzylpiperidine
A1086-25G
A1086-100G
TCA1086-025GEA
138.88
CAD
TCA1086-025G
TCA1086-500G
TCA1086-100G
4-Amino-1-benzylpiperidine ≥98.0%
4-Amino-1-benzylpiperidine
CAS Number: 50541-93-0
MDL Number: MFCD00006504
Molecular Formula: C12H18N2
Molecular Weight: 190.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.01
Formula:
C₁₂H₁₈N₂ MW: 190.29 g/mol Density: 1.01 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00006504 CAS Number: 50541-93-0 EINECS: 256-620-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...