Benzyl acetoacetate ≥95.0%

Supplier: TCI America

A1080-500ML A1080-25ML A1080-100ML
TCA1080-500MLEA 445.56 CAD
TCA1080-500ML TCA1080-025ML TCA1080-100ML
Benzyl acetoacetate ≥95.0%
Benzyl acetoacetate

CAS Number: 5396-89-4
MDL Number: MFCD00008785
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 163
Flash Point (°C): 108
Specific Gravity (20/20): 1.11


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Acetoacetic Acid Benzyl Ester

Formula: C₁₁H₁₂O₃
Boiling Pt: 163 °C
Density: 1.11 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00008785
CAS Number: 5396-89-4
EINECS: 226-416-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62496c3817cdc6c4effd7f0d259b26c1f36b6f25&vid=62496c3817cdc6c4effd7f0d259b26c1f36b6f25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR