3-(4-Aminophenyl)propionic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-Aminohydrocinnamic acid, 3-(4-Aminophenyl)propanoic acid

A1073-10G A1073-25G
TCA1073-010GEA 219.78 CAD
TCA1073-010G TCA1073-25G
3-(4-Aminophenyl)propionic acid ≥98.0%
3-(4-Aminophenyl)propionic acid

CAS Number: 2393-17-1
MDL Number: MFCD00017118
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 132


Other Product Names / Synonyms: 3-(4-Aminophenyl)propionic Acid

Formula: C₉H₁₁NO₂
Melting Pt: 132 °C
MDL Number: MFCD00017118
CAS Number: 2393-17-1
EINECS: 219-245-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07ac23ac38ba3b04dbfd44c14c0e88259f6c9a6d&vid=07ac23ac38ba3b04dbfd44c14c0e88259f6c9a6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR