(Aminomethyl)phosphonic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A1072-1G A1072-5G
TCA1072-001GEA 249.64 CAD
TCA1072-001G TCA1072-5G
(Aminomethyl)phosphonic acid ≥97.0% (by titrimetric analysis)
(Aminomethyl)phosphonic acid

CAS Number: 1066-51-9
MDL Number: MFCD00008105
Molecular Formula: CH6NO3P
Molecular Weight: 111.04
Purity/Analysis Method: >97.0% (T)
Form: Crystal

Formula: CH₆NO₃P
MDL Number: MFCD00008105
CAS Number: 1066-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dc38bdb8bdbe69069afce3e98993dff9a9ff2bc&vid=6dc38bdb8bdbe69069afce3e98993dff9a9ff2bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR