m-Anisic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-Anisic acid, 3-Methoxybenzoic acid

A1070-250G A1070-25G
TCA1070-250GEA 406.06 CAD
TCA1070-250G TCA1070-025G
m-Anisic acid ≥98.0%
m-Anisic acid

CAS Number: 586-38-9
MDL Number: MFCD00002499
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 172
Melting point (°C): 107


Other Product Names / Synonyms: 3-Methoxybenzoic Acid

Formula: C₈H₈O₃
Boiling Pt: 172 °C
Melting Pt: 107 °C
Density: 1.14 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002499
CAS Number: 586-38-9
EINECS: 209-574-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=414ee0bbdde364e28494d44deed7d0d969d004e9&vid=c90c6d01fb6a01d85cb162e25276ffbfcca4be2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR