2-Acetylfluorene ≥98.0%

Supplier: TCI America

Synonyms: Fluoren-2-yl methyl ketone

A1069-5G A1069-25G
TCA1069-5GEA 42.66 CAD
TCA1069-5G TCA1069-025G
2-Acetylfluorene ≥98.0%
2-Acetylfluorene

CAS Number: 781-73-7
MDL Number: MFCD00001124
Molecular Formula: C15H12O
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 130

Formula: C₁₅H₁₂O
Boiling Pt: 172…173 °C (0.05 mmHg)
Melting Pt: 130 °C
MDL Number: MFCD00001124
CAS Number: 781-73-7
EINECS: 212-310-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc14bad08effab333843f964e882cf0f8f0135f9&vid=dc14bad08effab333843f964e882cf0f8f0135f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR