1-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1068-25G A1068-100G
TCA1068-025GEA 317.58 CAD
TCA1068-025G TCA1068-100G
1-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-Aminocyclohexanecarboxylic acid

CAS Number: 2756-85-6
MDL Number: MFCD00001487
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₇H₁₃NO₂
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 170 °C
Density: 1.229 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001487
CAS Number: 2756-85-6
EINECS: 220-411-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7397972b389cc17f098502d2013cf0b8c34530e&vid=14af5f6c8f0da4506c76c49f564ee0daa73f18d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR