4-Chloro-2-(trifluoromethyl)aniline ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-5-chloro-α,α,α-trifluorotoluene, 2-Amino-5-chloro-α,α,α-trifluortoluene, 2-Amino-5-chlorobenzotrifluoride, 4-Chloro-α,α,α-trifluoro-o-toluidine

A1058-500G A1058-25G
TCA1058-500GEA 297.04 CAD
TCA1058-500G TCA1058-025G
4-Chloro-2-(trifluoromethyl)aniline ≥98.0%
4-Chloro-2-(trifluoromethyl)aniline

CAS Number: 445-03-4
MDL Number: MFCD00007841
Molecular Formula: C7H5ClF3N
Molecular Weight: 195.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 77
Flash Point (°C): 95
Specific Gravity (20/20): 1.44


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Chloro-2-trifluoromethylaniline

Formula: C₇H₅ClF₃N
Boiling Pt: 77 °C
Melting Pt: 9 °C
Density: 1.44 (20 °C)
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007841
CAS Number: 445-03-4
EINECS: 207-151-3
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9edb4c0a2eb68d9d0056b7e91c3c3c47a1f85a0d&vid=9edb4c0a2eb68d9d0056b7e91c3c3c47a1f85a0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR