4-Bromo-2-(trifluoromethyl)aniline ≥97.0%

Supplier: TCI America

Synonyms: 4-Bromo-α,α,α-trifluoro-o-toluidine, 2-Amino-5-bromobenzotrifluoride, 2-Amino-5-bromo-α,α,α-trifluorotoluene, 2-Amino-5-bromo-α,α,α-trifluortoluene

A1056-25G A1056-5G
TCA1056-25GEA 201.61 CAD
TCA1056-25G TCA1056-005G
4-Bromo-2-(trifluoromethyl)aniline ≥97.0%
4-Bromo-2-(trifluoromethyl)aniline

CAS Number: 445-02-3
MDL Number: MFCD00064393
Molecular Formula: C7H5BrF3N
Molecular Weight: 240.02
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 107
Specific Gravity (20/20): 1.72


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Bromo-2-trifluoromethylaniline

Formula: C₇H₅BrF₃N
Boiling Pt: 102 °C
Density: 1.72 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00064393
CAS Number: 445-02-3
EINECS: 207-150-8
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18afe440f99c5addd2926d3726d039f168ccc8af&vid=4eca19abfe18b4bf97d0c8f0e0bb028812635401&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR