1-Acetylnaphthalene ≥95.0%

Supplier: TCI America

Synonyms: 1'-Acetonaphthone, α-acetonaphthone, 1'-Acetylnaphthalene, Methyl α-naphthyl ketone, Methyl 1-naphthyl ketone, 1-Acetonaphthone

A1044-500G A1044-25G
TCA1044-500GEA 211.72 CAD
TCA1044-500G TCA1044-025G
1-Acetylnaphthalene ≥95.0%
1-Acetylnaphthalene

CAS Number: 941-98-0
MDL Number: MFCD00004013
Molecular Formula: C12H10O
Molecular Weight: 170.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 298
Flash Point (°C): 110
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 1-Acetylnaphthalene, Methyl 1-Naphthyl Ketone

Formula: C₁₂H₁₀O
Boiling Pt: 298 °C
Melting Pt: 9…11 °C
Density: 1.12 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004013
CAS Number: 941-98-0
EINECS: 213-384-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b3a625e5dc0997debceba4a19819fc0ceb8caf2&vid=7b3a625e5dc0997debceba4a19819fc0ceb8caf2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR