Methyl-3-aminothiophene-2-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3-amino-2-thiophenecarboxylate

A1039-25G A1039-100G
TCA1039-025GEA 82.16 CAD
TCA1039-025G TCA1039-100G
Methyl-3-aminothiophene-2-carboxylate ≥98.0%
Methyl 3-aminothiophene-2-carboxylate

CAS Number: 22288-78-4
MDL Number: MFCD00009765
Molecular Formula: C6H7NO2S
Molecular Weight: 157.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 102
Melting point (°C): 65
Flash Point (°C): 100


Other Product Names / Synonyms: 3-Amino-2-thiophenecarboxylic Acid Methyl Ester

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Boiling Pt: 102 °C
Melting Pt: 65 °C
Flash Pt: 100 °C
MDL Number: MFCD00009765
CAS Number: 22288-78-4
EINECS: 244-895-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0402db33a18e5808a63ab82dd51ffa4e845db5f1&vid=0402db33a18e5808a63ab82dd51ffa4e845db5f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR