D-(-)-Arginine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
A1029-5G
A1029-1G
TCA1029-5GEA
311.26
CAD
TCA1029-5G
TCA1029-001G
D-(-)-Arginine ≥98.0% (by titrimetric analysis)
D(-)-Arginine
CAS Number: 157-06-2
MDL Number: MFCD00063116
Molecular Formula: C6H14N4O2
Molecular Weight: 174.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -26 deg (C=8, 6mol/L HCl)
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Arg-OH
Formula:
C₆H₁₄N₄O₂ Melting Pt: 212 °C |
MDL Number:
MFCD00063116 CAS Number: 157-06-2 EINECS: 205-866-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...