4,4'-Dimethoxybenzil ≥98.0%

Supplier: TCI America

A1028-25G A1028-5G
TCA1028-025GEA 247.59 CAD
TCA1028-025G TCA1028-5G
4,4'-Dimethoxybenzil ≥98.0%
4,4'-Dimethoxybenzil

CAS Number: 1226-42-2
MDL Number: MFCD00008405
Molecular Formula: C16H14O4
Molecular Weight: 270.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 133


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 4,4'-Dimethoxybenzil

Formula: C₁₆H₁₄O₄
Boiling Pt: 215…219 °C (3 mmHg)
Melting Pt: 133 °C
Density: 1.36 g/cm³ (20 °C)
MDL Number: MFCD00008405
CAS Number: 1226-42-2
EINECS: 214-960-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22d377db000010e2f7650bb0089f95d43774eb6b&vid=22d377db000010e2f7650bb0089f95d43774eb6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR