6-Aminohexan-1-ol ≥97.0%

Supplier: TCI America
Warning

Synonyms: 6-Amino-1-hexanol

A1027-5G A1027-25G
TCA1027-005GEA 83.74 CAD
TCA1027-005G TCA1027-025G
6-Aminohexan-1-ol ≥97.0%
6-Aminohexan-1-ol

CAS Number: 4048-33-3
MDL Number: MFCD00008241
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 58

Formula: C₆H₁₅NO
Boiling Pt: 225 °C
Melting Pt: 58 °C
MDL Number: MFCD00008241
CAS Number: 4048-33-3
EINECS: 223-748-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cac594bdc29c3d113ecc2c51eac03a6b3bee19d&vid=1cac594bdc29c3d113ecc2c51eac03a6b3bee19d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR