4'-Phenylacetophenone ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
4-Acetylbiphenyl, 1-([1,1'-Biphenyl]-4-yl)ethanone, 4-Biphenylyl methyl ketone, 4'-Phenylacetophenone, p-Phenylacetophenone, 4-Acetyl-biphenyl
A1025-250G
A1025-25G
TCA1025-250GEA
257.86
CAD
TCA1025-250G
TCA1025-025G
4'-Phenylacetophenone ≥98.0%
4'-Phenylacetophenone
CAS Number: 92-91-1
MDL Number: MFCD00008749
Molecular Formula: C14H12O
Molecular Weight: 196.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 168
Melting point (°C): 122
Other Product Names / Synonyms: 4'-Phenylacetophenone
Formula:
C₁₄H₁₂O MW: 196.25 g/mol Boiling Pt: 325 Melting Pt: 122 °C Density: 1.251 g/cm³ (0 °C) |
MDL Number:
MFCD00008749 CAS Number: 92-91-1 EINECS: 202-202-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...