4-Acetylbenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A1024-1G
A1024-5G
TCA1024-001GEA
60.04
CAD
TCA1024-001G
TCA1024-005G
4-Acetylbenzoic acid ≥98.0% (by titrimetric analysis)
4-Acetylbenzoic acid
CAS Number: 586-89-0
MDL Number: MFCD00002561
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 210
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4'-Acetophenonecarboxylic Acid
Formula:
C₉H₈O₃ Boiling Pt: 340 °C (1013 hPa) Melting Pt: 210 °C |
MDL Number:
MFCD00002561 CAS Number: 586-89-0 EINECS: 209-588-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...