2-Acetyl-1-methyl-1H-pyrrole ≥98.0%

Supplier: TCI America

Synonyms: 1-(1-Methyl-1H-pyrrol-2-yl)ethanone, 2-Acetyl-1-methylpyrrole

A1014-5ML A1014-25ML
TCA1014-5MLEA 58.46 CAD
TCA1014-5ML TCA1014-025ML
2-Acetyl-1-methyl-1H-pyrrole ≥98.0%
2-Acetyl-1-methyl-1H-pyrrole

CAS Number: 932-16-1
MDL Number: MFCD00003089
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 202
Flash Point (°C): 82
Specific Gravity (20/20): 1.05

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 202 °C
Density: 1.05 (20 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003089
CAS Number: 932-16-1
EINECS: 213-247-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57cc5bf05b57eb180a629b0b5efab029baa8787f&vid=57cc5bf05b57eb180a629b0b5efab029baa8787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR