6-Methylanthranil acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Amino-6-methylbenzoic acid

A0996-5G A0996-25G
TCA0996-005GEA 97.96 CAD
TCA0996-005G TCA0996-025G
6-Methylanthranil acid ≥98.0%
6-Methylanthranil acid

CAS Number: 4389-50-8
MDL Number: MFCD00007809
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 124


Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 6-Amino-o-toluic Acid, 6-Methylanthranilic Acid

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 315
Melting Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00007809
CAS Number: 4389-50-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c39e330ecf3884f98d886bf16c59678a1b0e073&vid=d87f9044eccd34a72376e6e046e3394c94dbb22c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR