2-Aminopyrazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Aminopyrazine, Pyrazin-2-amine

A0989-5G A0989-25G
TCA0989-005GEA 58.46 CAD
TCA0989-005G TCA0989-025G
2-Aminopyrazine ≥98.0% (by GC, titration analysis)
2-Aminopyrazine

CAS Number: 5049-61-6
MDL Number: MFCD00006137
Molecular Formula: C4H5N3
Molecular Weight: 95.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 121

Formula: C₄H₅N₃
MW: 95.1 g/mol
Boiling Pt: 203
Melting Pt: 121 °C
MDL Number: MFCD00006137
CAS Number: 5049-61-6
EINECS: 225-748-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c0445f13bdae689ec317c45b17175d96cead041&vid=7c0445f13bdae689ec317c45b17175d96cead041&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR