2-Acetylpyrazine ≥99.0%

Supplier: TCI America

A0988-25G A0988-10G
TCA0988-25GEA 269.39 CAD
TCA0988-25G TCA0988-010G
2-Acetylpyrazine ≥99.0%
2-Acetylpyrazine

CAS Number: 22047-25-2
MDL Number: MFCD00006134
Molecular Formula: C6H6N2O
Molecular Weight: 122.13
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 72
Melting point (°C): 77

Formula: C₆H₆N₂O
Boiling Pt: 72 °C
Melting Pt: 77 °C
MDL Number: MFCD00006134
CAS Number: 22047-25-2
EINECS: 244-753-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb7070a0ea598274b7e2efb2780aa9081bc4b29c&vid=fb7070a0ea598274b7e2efb2780aa9081bc4b29c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR