4-Amino-2,3,5,6-tetrafluorobenzoic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 4-Amino-2,3,5,6-tetrafluorobenzoic Acid

A0987-5G A0987-1G
TCA0987-005GEA 467.68 CAD
TCA0987-005G TCA0987-1G
4-Amino-2,3,5,6-tetrafluorobenzoic acid ≥97.0% (by titrimetric analysis)
4-Amino-2,3,5,6-tetrafluorobenzoic acid

CAS Number: 944-43-4
MDL Number: MFCD00007647
Molecular Formula: C7H3F4NO2
Molecular Weight: 209.10
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 181

Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Melting Pt: 181 °C
MDL Number: MFCD00007647
CAS Number: 944-43-4
EINECS: 213-409-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=545e3355b34e0a1c65cf7d3ebae4c74c7a83721f&vid=6d42a6569872c0024c19d24d09a0f8d869ba4246&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR