4-Aminophthalimide ≥98.0%

Supplier: TCI America

A0966-5G A0966-25G
TCA0966-005GEA 116.92 CAD
TCA0966-005G TCA0966-025G
4-Aminophthalimide ≥98.0%
4-Aminophthalimide

CAS Number: 3676-85-5
MDL Number: MFCD00041854
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Formula: C₈H₆N₂O₂
Boiling Pt: 447 °C (1013 hPa)
Melting Pt: 294 °C
MDL Number: MFCD00041854
CAS Number: 3676-85-5
EINECS: 222-948-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29e60903f3c84ea83fe6d3594fcafa7c45d9443e&vid=29e60903f3c84ea83fe6d3594fcafa7c45d9443e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR