4-(Aminomethyl)benzoic acid ≥97.0%
Supplier: TCI America
A0965-25G
A0965-500G
A0965-100G
TCA0965-025GEA
67.94
CAD
TCA0965-025G
TCA0965-500G
TCA0965-100G
4-(Aminomethyl)benzoic acid ≥97.0%
4-(Aminomethyl)benzoic acid
CAS Number: 56-91-7
MDL Number: MFCD00010203
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Formula:
C₈H₉NO₂ MW: 151.16 g/mol |
MDL Number:
MFCD00010203 CAS Number: 56-91-7 EINECS: 200-297-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...