5-Aminoquinoline ≥99.0% (by titrimetric analysis), crystals

Supplier: TCI America
Warning

A0959-5G A0959-25G
TCA0959-5GEA 64.78 CAD
TCA0959-5G TCA0959-25G
5-Aminoquinoline ≥99.0% (by titrimetric analysis), crystals
5-Aminoquinoline

CAS Number: 611-34-7 MDL Number: MFCD00006797 Molecular Formula: C9H8N2 Molecular Weight: 144.18 Purity/Analysis Method: 99.0% (T) Form: Crystal Boiling point (°C): 310 Melting point (°C): 109

Formula: C₉H₈N₂
Boiling Pt: 310 °C
Melting Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00006797
CAS Number: 611-34-7
EINECS: 210-266-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12851bd610b5d634784b67983042bf23ec6df79a&vid=9ab166995544e4ee138f7a9d0c2b4c793d176327&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR