2-Amino-4-tert-butylphenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A0957-250G
A0957-25G
TCA0957-250GEA
580.18
CAD
TCA0957-250G
TCA0957-025G
2-Amino-4-tert-butylphenol ≥98.0% (by GC, titration analysis)
2-Amino-4-tert-butylphenol
CAS Number: 1199-46-8
MDL Number: MFCD00007698
Molecular Formula: C10H15NO
Molecular Weight: 165.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 162
Formula:
C₁₀H₁₅NO Melting Pt: 162 °C |
MDL Number:
MFCD00007698 CAS Number: 1199-46-8 EINECS: 214-844-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...