4-Amino-2-chlorobenzoic acid ≥97.0%

Supplier: TCI America
Warning

A0951-25G A0951-5G
TCA0951-25GEA 256.28 CAD
TCA0951-25G TCA0951-005G
4-Amino-2-chlorobenzoic acid ≥97.0%
4-Amino-2-chlorobenzoic acid

CAS Number: 2457-76-3
MDL Number: MFCD00007772
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 211

Formula: C₇H₆ClNO₂
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 211 °C
Storage Temperature: Ambient
MDL Number: MFCD00007772
CAS Number: 2457-76-3
EINECS: 219-540-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c68d60bbd24758daa710fc1c28e531b1329e829a&vid=f4c9e37ce997a32721daf4e263fc4db8c88cb17f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR