2-Amino-4-methoxy-6-methylpyrimidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0950-25G A0950-500G
TCA0950-025GEA 53.72 CAD
TCA0950-025G TCA0950-500G
2-Amino-4-methoxy-6-methylpyrimidine ≥98.0% (by titrimetric analysis)
2-Amino-4-methoxy-6-methylpyrimidine

CAS Number: 7749-47-5
MDL Number: MFCD00059768
Molecular Formula: C6H9N3O
Molecular Weight: 139.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 158


Other Product Names / Synonyms: 2-Amino-4-methyl-6-methoxypyrimidine

Formula: C₆H₉N₃O
Melting Pt: 158 °C
MDL Number: MFCD00059768
CAS Number: 7749-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1dba5cb77a6a70f174bf345d0f8a2ad51e0d6eb&vid=c1dba5cb77a6a70f174bf345d0f8a2ad51e0d6eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR