3-Aminocrotononitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

A0944-25G A0944-100G
TCA0944-025GEA 72.68 CAD
TCA0944-025G TCA0944-100G
3-Aminocrotononitrile ≥98.0% (by GC)
3-Aminocrotononitrile (mixture of cis and trans isomers)

Form: Crystal
Boiling point (°C): 120
Melting point (°C): 75
Flash Point (°C): 154


Other Product Names / Synonyms: 3-Amino-2-butenonitrile, Diacetonitrile

Formula: C₄H₆N₂
Boiling Pt: 120 °C
Melting Pt: 75 °C
Flash Pt: 154 °C
MDL Number: MFCD00008071
CAS Number: 1118-61-2
EINECS: 214-266-2
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9b54ba6a86f7e7f46b712a4e9f030cc4d992404&vid=c9b54ba6a86f7e7f46b712a4e9f030cc4d992404&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR