1-Adamantanol ≥98.0%

Supplier: TCI America

Synonyms: 1-Tricyclo[3.3.1.1(3,7)]decanol, 1-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-1-ol, Adamantan-1-ol

A0939-25G A0939-250G
TCA0939-025GEA 127.35 CAD
TCA0939-025G TCA0939-250G
1-Adamantanol ≥98.0%
1-Adamantanol

CAS Number: 768-95-6
MDL Number: MFCD00074729
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 280


Other Product Names / Synonyms: 1-Hydroxyadamantane

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 280 °C
Density: 1.06 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074729
CAS Number: 768-95-6
EINECS: 212-202-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5216d4617671caaa06c7139fb81dc63f1d02f92a&vid=5216d4617671caaa06c7139fb81dc63f1d02f92a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR