Ethyl-p-anisate

Supplier: TCI America

A0933-25ML A0933-100ML
TCA0933-025MLEA 65.89 CAD
TCA0933-025ML TCA0933-500ML TCA0933-100ML
Ethyl-p-anisate
Ethyl p-anisate

CAS Number: 94-30-4
MDL Number: MFCD00015151
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 143
Flash Point (°C): 138
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: p-Anisic Acid Ethyl Ester, Ethyl 4-Methoxybenzoate, 4-Methoxybenzoic Acid Ethyl Ester

Formula: C₁₀H₁₂O₃
Boiling Pt: 143 °C
Density: 1.11 (20 °C)
Flash Pt: 138 °C
MDL Number: MFCD00015151
CAS Number: 94-30-4
EINECS: 202-320-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7080952081ae5c9b95e505e341b6cacdcf93126d&vid=7080952081ae5c9b95e505e341b6cacdcf93126d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR