12-Aminolauric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0932-25G A0932-500G
TCA0932-025GEA 68.11 CAD
TCA0932-025G TCA0932-500G
12-Aminolauric acid ≥98.0% (by titrimetric analysis)
12-Aminolauric acid

CAS Number: 693-57-2
MDL Number: MFCD00008153
Molecular Formula: C12H25NO2
Molecular Weight: 215.34
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 186
Flash Point (°C): 190


Other Product Names / Synonyms: 12-Aminododecanoic Acid

Formula: C₁₂H₂₅NO₂
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 186 °C
Flash Pt: 190 °C
MDL Number: MFCD00008153
CAS Number: 693-57-2
EINECS: 211-754-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a6bb8a0688876febbf263167ae0d4b8f8dee9b2&vid=5a6bb8a0688876febbf263167ae0d4b8f8dee9b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR